5-Cyanothiophene-2-boronic acid, pinacol ester 96%

BLDPBD162299-5GEA 182 EUR
BLDPBD162299-5G BLDPBD162299-250MG BLDPBD162299-1G
5-Cyanothiophene-2-boronic acid, pinacol ester 96%
5-Cyanothiophene-2-boronic acid, pinacol ester

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₁H₁₄BNO₂S
Molekulargewicht: 235,11 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09952061
CAS-Nummer: 676501-85-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR