4-Butylbiphenyl 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD16401-25GEA 24 EUR
BLDPBD16401-25G BLDPBD16401-10G
4-Butylbiphenyl 98%
4-Butylbiphenyl

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₆H₁₈
Molekulargewicht: 210,32 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00100665
CAS-Nummer: 37909-95-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR