3-Brompyridazin 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD164106-10GEA 244 EUR
BLDPBD164106-10G BLDPBD164106-5G BLDPBD164106-1G BLDPBD164106-250MG
3-Brompyridazin 97%
3-Bromopyridazine

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₄H₃BrN₂
Molekulargewicht: 158,9840 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD10688587
CAS-Nummer: 88491-61-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR