7-Bromo-2,3-dihydrobenzofuran 98%

BLDPBD164145-250MGEA 19 EUR
BLDPBD164145-250MG BLDPBD164145-5G BLDPBD164145-100MG BLDPBD164145-1G
7-Bromo-2,3-dihydrobenzofuran 98%
7-Bromo-2,3-dihydrobenzofuran

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₈H₇BrO
Molekulargewicht: 199,045 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD09266210
CAS-Nummer: 206347-30-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR