Methylimidazo[1,2-a]pyridin-7-carboxylat 95%
|
|
BLDPBD164815-5GEA
128
EUR
BLDPBD164815-5G
BLDPBD164815-250MG
BLDPBD164815-1G
BLDPBD164815-10G
Methylimidazo[1,2-a]pyridin-7-carboxylat 95%
Methylimidazo[1,2-a]pyridin-7-carboxylat
Storage condition: Sealed in dry, 2-8°C
Formel:
C₉H₈N₂O₂ Molekulargewicht: 176,18 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD09910350 CAS-Nummer: 86718-01-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...