[6-[(tert-Butoxycarbonyl)amino]pyridin-3-yl]boronic acid 98%

BLDPBD164823-100MGEA 22 EUR
BLDPBD164823-100MG BLDPBD164823-1G BLDPBD164823-5G BLDPBD164823-250MG
[6-[(tert-Butoxycarbonyl)amino]pyridin-3-yl]boronic acid 98%
[6-[(tert-Butoxycarbonyl)amino]pyridin-3-yl]boronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₁₅BN₂O₄
Molekulargewicht: 238,048 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD11975411
CAS-Nummer: 883231-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR