4-Piperazinylphenylboronic acid, pinacol ester 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD164843-1GEA 121 EUR
BLDPBD164843-1G BLDPBD164843-100MG BLDPBD164843-5G BLDPBD164843-250MG
4-Piperazinylphenylboronic acid, pinacol ester 95%
4-Piperazinylphenylboronic acid, pinacol ester

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₆H₂₅BN₂O₂
Molekulargewicht: 288,193 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD06795649
CAS-Nummer: 912369-50-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR