1H-Indazol-6-boronsäurepinakolester 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD164851-100MGEA 26 EUR
BLDPBD164851-100MG BLDPBD164851-5G BLDPBD164851-1G BLDPBD164851-250MG BLDPBD164851-10G
1H-Indazol-6-boronsäurepinakolester 98%
1H-Indazol-6-boronsäurepinakolester

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₃H₁₇BN₂O₂
Molekulargewicht: 244,0973 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD07368067
CAS-Nummer: 937049-58-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR