1H-Indazol-5-carbonsäure 98%
|
|
BLDPBD165362-1GEA
11
EUR
BLDPBD165362-1G
BLDPBD165362-25G
BLDPBD165362-250MG
BLDPBD165362-5G
BLDPBD165362-10G
1H-Indazol-5-carbonsäure 98%
1H-Indazole-5-carboxylic acid
Storage condition: Sealed in dry, Room Temperature
Formel:
C₈H₆N₂O₂ Molekulargewicht: 162,15 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06804570 CAS-Nummer: 61700-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...