4-(2-Pyridinyl)phenylboronic acid, pinacol ester 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD165818-250MGEA 15 EUR
BLDPBD165818-250MG BLDPBD165818-1G BLDPBD165818-100MG
4-(2-Pyridinyl)phenylboronic acid, pinacol ester 95%
4-(2-Pyridinyl)phenylboronic acid, pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₇H₂₀BNO₂
Molekulargewicht: 281,157 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD11973624
CAS-Nummer: 908350-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR