[2-(1,3-Dioxan-2-yl)ethyl]triphenylphosphoniumbromid 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD1672-5GEA 46 EUR
BLDPBD1672-5G BLDPBD1672-1G BLDPBD1672-25G BLDPBD1672-100G
[2-(1,3-Dioxan-2-yl)ethyl]triphenylphosphoniumbromid 98%
[2-(1,3-Dioxan-2-yl)ethyl]triphenylphosphoniumbromid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₂₄H₂₆BrO₂P
Molekulargewicht: 457,36 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00012000
CAS-Nummer: 69891-92-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR