6-Bromo-2,3-diaminopyridine 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD167473-1GEA 116 EUR
BLDPBD167473-1G BLDPBD167473-5G BLDPBD167473-250MG BLDPBD167473-10G BLDPBD167473-100MG BLDPBD167473-25G
6-Bromo-2,3-diaminopyridine 97%
6-Bromo-2,3-diaminopyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₅H₆BrN₃
Molekulargewicht: 188,025 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD10697695
CAS-Nummer: 129012-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR