2-Methyl-5-(pyrrolidin-2-yl)pyridine 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD167687-250MGEA 190 EUR
BLDPBD167687-250MG BLDPBD167687-1G BLDPBD167687-100MG
2-Methyl-5-(pyrrolidin-2-yl)pyridine 97%
2-Methyl-5-(pyrrolidin-2-yl)pyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₀H₁₄N₂
Molekulargewicht: 162,232 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD12024458
CAS-Nummer: 90872-72-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR