2-Bromo-N-phenyl-acetamide 97%

BLDPBD168139-250MGEA 40 EUR
BLDPBD168139-250MG BLDPBD168139-1G BLDPBD168139-100MG BLDPBD168139-5G
2-Bromo-N-phenyl-acetamide 97%
2-Bromo-N-phenyl-acetamide

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₈BrNO
Molekulargewicht: 214,059 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00462518
CAS-Nummer: 5326-87-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR