4-Cyan-4'-pentyloxybiphenyl 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD16887-10GEA 81 EUR
BLDPBD16887-10G BLDPBD16887-25G BLDPBD16887-5G BLDPBD16887-1G
4-Cyan-4'-pentyloxybiphenyl 98%
4-Cyan-4'-pentyloxybiphenyl

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₈H₁₉NO
Molekulargewicht: 281,42 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00074879
CAS-Nummer: 52364-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR