4-Methoxyzimtsäure 98%

BLDPBD17019-100GEA 21 EUR
BLDPBD17019-100G BLDPBD17019-10G BLDPBD17019-5G BLDPBD17019-500G BLDPBD17019-25G BLDPBD17019-1000G
4-Methoxyzimtsäure 98%
4-Methoxyzimtsäure

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₀H₁₀O₃
Molekulargewicht: 178,18 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00004398
CAS-Nummer: 830-09-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR