2-Methyl-4-(1H-pyrazol-1-yl)benzaldehyd 98%

BLDPBD171014-1GEA 45 EUR
BLDPBD171014-1G BLDPBD171014-25G BLDPBD171014-5G
2-Methyl-4-(1H-pyrazol-1-yl)benzaldehyd 98%
2-Methyl-4-(1H-pyrazol-1-yl)benzaldehyd

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₁H₁₀N₂O
Molekulargewicht: 186,2099 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD08059904
CAS-Nummer: 1015845-89-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR