HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure) 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD17277-25GEA 7 EUR
BLDPBD17277-25G BLDPBD17277-1000G BLDPBD17277-100G BLDPBD17277-0.5G BLDPBD17277-500G
HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure) 97%
HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure)

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₁₈N₂O₄S
Molekulargewicht: 238,31 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00006158
CAS-Nummer: 7365-45-9
https://at.vwr.com/stibo/web/std.lang.all/38/07/41813807.jpg

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR