2,4-Dichlor-L-phenylalanin 97%

BLDPBD17520-5GEA 48 EUR
BLDPBD17520-5G BLDPBD17520-1G BLDPBD17520-25G
2,4-Dichlor-L-phenylalanin 97%
2,4-Dichlor-L-phenylalanin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₉Cl₂NO₂
Molekulargewicht: 234,1 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01860882
CAS-Nummer: 111119-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR