2-Ethoxy-5-nitrobenzonitril 97%

BLDPBD178057-5GEA 117 EUR
BLDPBD178057-5G BLDPBD178057-250MG BLDPBD178057-1G
2-Ethoxy-5-nitrobenzonitril 97%
2-Ethoxy-5-nitrobenzonitril

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₈N₂O₃
Molekulargewicht: 192,1700 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD02256084
CAS-Nummer: 29669-41-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR