3-Chlor-6-(propylamino)pyridazin 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD180455-25GEA 545 EUR
BLDPBD180455-25G BLDPBD180455-5G BLDPBD180455-1G
3-Chlor-6-(propylamino)pyridazin 98%
3-Chloro-6-(propylamino)pyridazine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₇H₁₀ClN₃
Molekulargewicht: 171,6274 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09800940
CAS-Nummer: 951885-19-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR