Phenacyltriphenylphosphoniumbromid 98%

BLDPBD1811-25GEA 56 EUR
BLDPBD1811-25G BLDPBD1811-5G BLDPBD1811-100G
Phenacyltriphenylphosphoniumbromid 98%
Phenacyltriphenylphosphoniumbromid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₂₆H₂₂BrOP
Molekulargewicht: 461,3301 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011920
CAS-Nummer: 6048-29-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR