Methyl-2-amino-4-brombenzoat 97%

BLDPBD18134-100GEA 189 EUR
BLDPBD18134-100G BLDPBD18134-25G BLDPBD18134-1G BLDPBD18134-10G BLDPBD18134-5G BLDPBD18134-500G
Methyl-2-amino-4-brombenzoat 97%
Methyl-2-amino-4-brombenzoat
Formel: C₈H₈BrNO₂
Molekulargewicht: 230,0586 g/mol
MDL Number: MFCD07783014
CAS-Nummer: 135484-83-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR