Methyl-4-bromsalicylat 98%

BLDPBD182298-1GEA 11 EUR
BLDPBD182298-1G BLDPBD182298-5G BLDPBD182298-10G BLDPBD182298-25G BLDPBD182298-250MG
Methyl-4-bromsalicylat 98%
Methyl-4-bromsalicylat

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₇BrO₃
Molekulargewicht: 231,0434 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07780736
CAS-Nummer: 22717-56-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR