Bis(benzonitril)dichlorpalladium(II) 99% Pd>27.7%

BLDPBD18299-5GEA 372 EUR
BLDPBD18299-5G BLDPBD18299-100MG BLDPBD18299-250MG BLDPBD18299-1G
Bis(benzonitril)dichlorpalladium(II) 99% Pd>27.7%
Bis(benzonitril)dichlorpalladium(II)

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₄H₁₀Cl₂N₂Pd
Molekulargewicht: 383,57 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00013123
CAS-Nummer: 14220-64-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR