1-Brom-3,5-difluor-4-iodbenzol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD18678-10GEA 25 EUR
BLDPBD18678-10G BLDPBD18678-5G BLDPBD18678-500G BLDPBD18678-0.5G BLDPBD18678-25G BLDPBD18678-100G
1-Brom-3,5-difluor-4-iodbenzol 98%
1-Bromo-3,5-difluoro-4-iodobenzene

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₂BrF₂I
Molekulargewicht: 318,8854 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01861126
CAS-Nummer: 160976-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR