4-Phenyl-3-butin-2-on 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD18974-25GEA 427 EUR
BLDPBD18974-25G BLDPBD18974-5G BLDPBD18974-1G
4-Phenyl-3-butin-2-on 97%
4-Phenyl-3-butin-2-on

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₀H₈O
Molekulargewicht: 144,17 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00008776
CAS-Nummer: 1817-57-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR