3-Chlor-4-formylphenylboronsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD196281-1GEA 241 EUR
BLDPBD196281-1G BLDPBD196281-100MG BLDPBD196281-250MG
3-Chlor-4-formylphenylboronsäure 98%
3-Chlor-4-formylphenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₆BClO₃
Molekulargewicht: 184,39 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD08274473
CAS-Nummer: 1072952-53-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR