2-Chlor-6-methylisonicotinsäure 98%
|
|
BLDPBD19716-25GEA
69
EUR
BLDPBD19716-25G
BLDPBD19716-10G
BLDPBD19716-5G
BLDPBD19716-1G
BLDPBD19716-100G
BLDPBD19716-500G
BLDPBD19716-250MG
2-Chlor-6-methylisonicotinsäure 98%
2-Chlor-6-methylisonicotinsäure
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₇H₆ClNO₂ Molekulargewicht: 171,58 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00052830 CAS-Nummer: 25462-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...