Benzyl-3-oxopiperidine-1-carboxylate 97%

BLDPBD1990-10GEA 46 EUR
BLDPBD1990-10G BLDPBD1990-5G BLDPBD1990-500G BLDPBD1990-1G BLDPBD1990-100G BLDPBD1990-25G
Benzyl-3-oxopiperidine-1-carboxylate 97%
Benzyl-3-oxopiperidine-1-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₃H₁₅NO₃
Molekulargewicht: 233,27 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD03001723
CAS-Nummer: 61995-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR