Methyl-3-iodimidazo[1,2-a]pyridin-6-carboxylat 98%

BLDPBD200022-1GEA 108 EUR
BLDPBD200022-1G BLDPBD200022-250MG BLDPBD200022-5G
Methyl-3-iodimidazo[1,2-a]pyridin-6-carboxylat 98%
Methyl-3-iodimidazo[1,2-a]pyridin-6-carboxylat

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₉H₇IN₂O₂
Molekulargewicht: 302,0686 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD07021385
CAS-Nummer: 460087-82-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR