D(+)-Glucuronsäure-6,3-lacton 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD20224-25GEA 7 EUR
BLDPBD20224-25G BLDPBD20224-500G BLDPBD20224-100G BLDPBD20224-10G BLDPBD20224-1000G
D(+)-Glucuronsäure-6,3-lacton 98%
D(+)-Glucuronsäure-6,3-lacton

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₆H₈O₆
Molekulargewicht: 176,12 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00135622
CAS-Nummer: 32449-92-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR