2-Chloro-3-chloromethyl-pyridine 98%

BLDPBD204371-5GEA 116 EUR
BLDPBD204371-5G BLDPBD204371-250MG BLDPBD204371-1G BLDPBD204371-10G
2-Chloro-3-chloromethyl-pyridine 98%
2-Chloro-3-chloromethyl-pyridine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₅Cl₂N
Molekulargewicht: 162,017 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09835107
CAS-Nummer: 89581-84-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR