5-Nitro-1,2,3-benzenetricarboxylic Acid 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD20559-10GEA 82 EUR
BLDPBD20559-10G BLDPBD20559-1G BLDPBD20559-25G BLDPBD20559-5G
5-Nitro-1,2,3-benzenetricarboxylic Acid 95%
5-Nitro-1,2,3-benzenetricarboxylic Acid
Formel: C₉H₅NO₈
Molekulargewicht: 255,138 g/mol
MDL Number: MFCD03839939
CAS-Nummer: 3807-81-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR