(R)-(+)-N-Benzyl-α-methylbenzylamin 96%
Lieferant: BLD PHARMATECH GMBH
|
|
BLDPBD20572-25GEA
24
EUR
BLDPBD20572-25G
BLDPBD20572-100G
BLDPBD20572-10G
BLDPBD20572-500G
BLDPBD20572-5G
(R)-(+)-N-Benzyl-α-methylbenzylamin 96%
(R)-(+)-N-Benzyl-α-methylbenzylamin
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₁₅H₁₇N Molekulargewicht: 211,31 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00015010 CAS-Nummer: 38235-77-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...