3-Fluor-2-iodbenzoesäure 98%

BLDPBD20588-10GEA 257 EUR
BLDPBD20588-10G BLDPBD20588-25G BLDPBD20588-1G BLDPBD20588-250MG BLDPBD20588-5G
3-Fluor-2-iodbenzoesäure 98%
3-Fluoro-2-iodobenzoic acid

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₇H₄FIO₂
Molekulargewicht: 266,0100 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD07774249
CAS-Nummer: 387-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR