2,6-Dibromo-4-methoxypyridine 96%

Lieferant: BLD PHARMATECH GMBH

BLDPBD208279-10GEA 378 EUR
BLDPBD208279-10G BLDPBD208279-250MG BLDPBD208279-5G BLDPBD208279-25G BLDPBD208279-1G
2,6-Dibromo-4-methoxypyridine 96%
2,6-Dibromo-4-methoxypyridine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₅Br₂NO
Molekulargewicht: 266,918 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD11036227
CAS-Nummer: 117873-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR