Methyl 3-bromo-5-methylbenzoate 97%

BLDPBD208315-100MGEA 6 EUR
BLDPBD208315-100MG BLDPBD208315-10G BLDPBD208315-5G BLDPBD208315-1G BLDPBD208315-250MG BLDPBD208315-25G
Methyl 3-bromo-5-methylbenzoate 97%
Methyl 3-bromo-5-methylbenzoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₉BrO₂
Molekulargewicht: 229,071 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11007827
CAS-Nummer: 478375-40-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR