4-Amino-3-methoxybenzonitril 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD208956-10GEA 392 EUR
BLDPBD208956-10G BLDPBD208956-1G BLDPBD208956-25G BLDPBD208956-5G BLDPBD208956-250MG BLDPBD208956-100MG
4-Amino-3-methoxybenzonitril 98%
4-Amino-3-methoxybenzonitril

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₈N₂O
Molekulargewicht: 148,1600 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD13250177
CAS-Nummer: 177476-76-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR