1,3-Dimethyl-1H-pyrazol-4-boronsäurepinakolester 97%

BLDPBD208999-250MGEA 7 EUR
BLDPBD208999-250MG BLDPBD208999-100G BLDPBD208999-10G BLDPBD208999-25G BLDPBD208999-1G BLDPBD208999-0.025 BLDPBD208999-5G
1,3-Dimethyl-1H-pyrazol-4-boronsäurepinakolester 97%
1,3-Dimethyl-1H-pyrazol-4-boronsäurepinakolester

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₁H₁₉BN₂O₂
Molekulargewicht: 222,0918 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09864190
CAS-Nummer: 1046832-21-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR