Ethyl 7-bromoimidazo[1,2-a]pyridine-3-carboxylate 97%

BLDPBD209041-1GEA 87 EUR
BLDPBD209041-1G BLDPBD209041-5G BLDPBD209041-250MG
Ethyl 7-bromoimidazo[1,2-a]pyridine-3-carboxylate 97%
Ethyl 7-bromoimidazo[1,2-a]pyridine-3-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₉BrN₂O₂
Molekulargewicht: 269,095 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11045695
CAS-Nummer: 1134327-98-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR