2-(Chloromethyl)-4,6-dimethylpyrimidine 97%

BLDPBD210742-5GEA 961 EUR
BLDPBD210742-5G BLDPBD210742-1G BLDPBD210742-100MG BLDPBD210742-250MG BLDPBD210742-10G
2-(Chloromethyl)-4,6-dimethylpyrimidine 97%
2-(Chloromethyl)-4,6-dimethylpyrimidine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₉ClN₂
Molekulargewicht: 156,613 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09864267
CAS-Nummer: 74502-83-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR