4-Pyridazincarbonsäure 97%

BLDPBD21076-25GEA 664 EUR
BLDPBD21076-25G BLDPBD21076-1G BLDPBD21076-250MG BLDPBD21076-5G BLDPBD21076-10G
4-Pyridazincarbonsäure 97%
Pyridazine-4-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₅H₄N₂O₂
Molekulargewicht: 124,10 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011576
CAS-Nummer: 50681-25-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR