2-Propylimidazol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD21103-100GEA 17 EUR
BLDPBD21103-100G BLDPBD21103-25G BLDPBD21103-500G BLDPBD21103-1000G BLDPBD21103-10G
2-Propylimidazol 98%
2-Propylimidazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₁₀N₂
Molekulargewicht: 110,1570 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00059158
CAS-Nummer: 50995-95-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR