Ethyl-5-methylisoxazol-4-carboxylat 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD21115-10GEA 73 EUR
BLDPBD21115-10G BLDPBD21115-1G BLDPBD21115-250MG BLDPBD21115-5G BLDPBD21115-25G
Ethyl-5-methylisoxazol-4-carboxylat 97%
Ethyl-5-methylisoxazol-4-carboxylat

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₉NO₃
Molekulargewicht: 155,15 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01631119
CAS-Nummer: 51135-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR