3-Nitro-2-pyridincarbonitril 98%

BLDPBD21128-5GEA 114 EUR
BLDPBD21128-5G BLDPBD21128-1G BLDPBD21128-250MG
3-Nitro-2-pyridincarbonitril 98%
3-Nitro-2-pyridincarbonitril

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₃N₃O₂
Molekulargewicht: 149,1100 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06658499
CAS-Nummer: 51315-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR