5-Carboxyfuran-2-boronic acid 96%

Lieferant: BLD PHARMATECH GMBH

BLDPBD211750-100MGEA 62 EUR
BLDPBD211750-100MG BLDPBD211750-250MG
5-Carboxyfuran-2-boronic acid 96%
5-Carboxyfuran-2-boronic acid

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₅H₅BO₅
Molekulargewicht: 155,901 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD04113807
CAS-Nummer: 852228-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR