3-Fluor-5-hydroxybenzonitril 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD211892-25GEA 117 EUR
BLDPBD211892-25G BLDPBD211892-1G BLDPBD211892-10G BLDPBD211892-5G BLDPBD211892-100G
3-Fluor-5-hydroxybenzonitril 97%
3-Fluor-5-hydroxybenzonitril

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₄FNO
Molekulargewicht: 137,1112 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09972133
CAS-Nummer: 473923-95-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR