3-Bromo-5-nitroaniline 95%

BLDPBD212777-25GEA 352 EUR
BLDPBD212777-25G BLDPBD212777-250MG BLDPBD212777-1G BLDPBD212777-5G
3-Bromo-5-nitroaniline 95%
3-Bromo-5-nitroaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₅BrN₂O₂
Molekulargewicht: 217,02 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00462820
CAS-Nummer: 55215-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR