(S)-(+)-Ethyl-3-piperidincarboxylat 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD212809-10GEA 34 EUR
BLDPBD212809-10G BLDPBD212809-5G BLDPBD212809-100G BLDPBD212809-25G
(S)-(+)-Ethyl-3-piperidincarboxylat 98%
(S)-(+)-Ethyl-3-piperidincarboxylat

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formel: C₈H₁₅NO₂
Molekulargewicht: 157,21 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD00792499
CAS-Nummer: 37675-18-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR